Gene Transcripts

Gene Symbol
CDSN
Entrez Gene ID
1041
Gene Name
corneodesmosin
Gene Aliases
HTSS, HTSS1, HYPT2, PSS, PSS1, S
Chromosome Location
Chr. 6: 31115088 - 31120475 on Build GRCh38
UniGene ID
Hs.310958
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001264.4 NP_001255.3 2 999
GenBank mRNA AF030130.1 - 2 957
AK292636.1 - 2 967
BC031993.1 - 2 947 4747362
L20815.1 - 2 957

View Full Product Details