Gene Transcripts
- Gene Symbol
- LPAR1
- Entrez Gene ID
- 1902
- Gene Name
- lysophosphatidic acid receptor 1
- Gene Aliases
- EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
- Chromosome Location
- Chr. 9: 110873252 - 111039259 on Build GRCh38
- UniGene ID
- Hs.126667
- Species
- Homo sapiens
View Full Product Details