Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001447.2 | NP_001438.1 | 1 | 1311 | |
XM_006714761.3 | XP_006714824.1 | 2 | 1882 | ||
XM_011537600.2 | XP_011535902.1 | 4 | 1650 | ||
XM_011537603.2 | XP_011535905.1 | 2 | 1583 | ||
XM_017009224.1 | XP_016864713.1 | 4 | 1652 | ||
XM_017009225.1 | XP_016864714.1 | 3 | 1494 | ||
XM_017009226.1 | XP_016864715.1 | 2 | 1887 | ||
XM_017009227.1 | XP_016864716.1 | 2 | 1886 | ||
GenBank mRNA | AF231022.1 | - | 1 | 1311 | |
AK308619.1 | - | 2 | 1476 |