Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001099735.1 | NP_001093205.1 | 5 | 557 | |
NM_001099736.1 | NP_001093206.1 | 5 | 561 | ||
NM_001825.2 | NP_001816.2 | 6 | 721 | ||
GenBank mRNA | AK122769.1 | - | 5 | 521 | |
AK313293.1 | - | 5 | 548 | ||
BC029140.1 | - | 5 | 538 | 5195322 | |
CR450315.1 | - | 4 | 479 | ||
DC308656.1 | - | 5 | 561 | ||
J05401.1 | - | 5 | 677 |