Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290307.2 | NP_001277236.1 | 3 | 248 | |
NM_001290310.2 | NP_001277239.1 | 3 | 248 | ||
NM_001323982.1 | NP_001310911.1 | 4 | 718 | ||
NM_001323983.1 | NP_001310912.1 | 3 | 400 | ||
NM_001323984.1 | NP_001310913.1 | 4 | 266 | ||
NM_001323985.1 | NP_001310914.1 | 3 | 248 | ||
NM_001323986.1 | NP_001310915.1 | 3 | 248 | ||
NM_001903.4 | NP_001894.2 | 3 | 248 | ||
GenBank mRNA | AK094151.1 | - | 3 | 187 | |
AK296708.1 | - | 3 | 219 | ||
AK304267.1 | - | 3 | 188 | ||
AU141929.2 | - | 3 | 216 | ||
BC000385.2 | - | 3 | 186 | 2821111 | |
D13866.1 | - | 3 | 187 | ||
D14705.1 | - | 3 | 212 | ||
DC332608.1 | - | 3 | 248 | ||
DC369291.1 | - | 3 | 238 | ||
L23805.1 | - | 2 | 125 | ||
U03100.1 | - | 2 | 125 |