Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001931.4 | NP_001922.2 | 8 | 1802 | |
XM_011542647.2 | XP_011540949.1 | 8 | 1449 | ||
GenBank mRNA | AK057299.1 | - | 8 | 1802 | |
AK223596.1 | - | 8 | 1191 | ||
AK296274.1 | - | 8 | 1081 | ||
AK297103.1 | - | 5 | 548 | ||
AK299562.1 | - | 6 | 639 | ||
AK303930.1 | - | 9 | 1027 | ||
BC039084.1 | - | 8 | 1171 | 4508489 | |
X13822.1 | - | 2 | 135 | ||
Y00978.1 | - | 8 | 1047 |