Gene Transcripts

Gene Symbol
FGD1
Entrez Gene ID
2245
Gene Name
FYVE, RhoGEF and PH domain containing 1
Gene Aliases
AAS, FGDY, MRXS16, ZFYVE3
Chromosome Location
Chr. X: 54445454 - 54496166 on Build GRCh38
UniGene ID
Hs.709201
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004463.2 NP_004454.2 4 1590
GenBank mRNA AK226117.1 - 4 1578
AK299635.1 - 2 299
BC034530.1 - 4 1102 4938457
U11690.1 - 4 1586

View Full Product Details