Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256295.1 | NP_001243224.1 | 10 | 1629 | |
NM_005239.5 | NP_005230.1 | 9 | 1459 | ||
XM_005260935.1 | XP_005260992.1 | 9 | 1532 | ||
XM_017028290.1 | XP_016883779.1 | 9 | 4129 | ||
GenBank mRNA | AK096841.1 | - | 10 | 1629 | |
AK315563.1 | - | 9 | 1362 | ||
BC017040.1 | - | 9 | 1331 | 3852274 | |
BC042954.2 | - | 9 | 1361 | 5276704 | |
BT006838.1 | - | 8 | 1169 | ||
J04102.1 | - | 9 | 1460 | ||
M11922.1 | - | 2 | 200 |