Gene Transcripts

Gene Symbol
EVI2B
Entrez Gene ID
2124
Gene Name
ecotropic viral integration site 2B
Gene Aliases
CD361, D17S376, EVDB
Chromosome Location
Chr. 17: 31303766 - 31314149 on Build GRCh38
UniGene ID
Hs.5509
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006495.3 NP_006486.3 2 253
XM_005257946.3 XP_005258003.1 3 363
GenBank mRNA AK297087.1 - 3 298
AK312822.1 - 2 198
BC005926.1 - 2 214 4043339
DA677356.1 - 2 254

View Full Product Details