Gene Transcripts

Gene Symbol
FANCF
Entrez Gene ID
2188
Gene Name
Fanconi anemia complementation group F
Gene Aliases
FAF
Chromosome Location
Chr. 11: 22622533 - 22625841 on Build GRCh38
UniGene ID
Hs.632151
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022725.3 NP_073562.1 1 709
GenBank mRNA AF181994.1 - 1 682
AF181995.1 - 1 678
AK001716.2 - 1 691
AK023153.1 - 1 691
AK223277.1 - 1 683
BC035346.1 - 1 703
BC047028.1 - 1 681 5538578
BC063038.1 - 1 709 5239725
BC093867.1 - 1 699 7939712
BC101807.1 - 1 699 8069313
BI522448.1 - 1 709
BX640996.1 - 1 701

View Full Product Details