Gene Transcripts

Gene Symbol
ADH6
Entrez Gene ID
130
Gene Name
alcohol dehydrogenase 6 (class V)
Gene Aliases
ADH-5
Chromosome Location
Chr. 4: 99202638 - 99219246 on Build GRCh38
UniGene ID
Hs.586161
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000672.3 NP_000663.1 8 1076
NM_001102470.1 NP_001095940.1 8 1076
NR_132990.1 6 717
GenBank mRNA AK092768.1 - 8 1076
AK298290.1 - 4 645
AK316326.1 - 6 645
BC039065.1 - 7 929 5184104

View Full Product Details