Gene Transcripts

Gene Symbol
ACR
Entrez Gene ID
49
Gene Name
acrosin
Gene Aliases
Chromosome Location
Chr. 22: 50738224 - 50745299 on Build GRCh38
UniGene ID
Hs.370870
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097.2 NP_001088.2 5 1216
GenBank mRNA CR456366.1 - 5 1211
Y00970.1 - 5 1220

View Full Product Details