Gene Transcripts

Gene Symbol
CRISP1
Entrez Gene ID
167
Gene Name
cysteine rich secretory protein 1
Gene Aliases
AEGL1, ARP, CRISP-1, HEL-S-57, HSCRISP1D, HSCRISP1G, HUMARP
Chromosome Location
Chr. 6: 49834257 - 49877096 on Build GRCh38
UniGene ID
Hs.109620
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001131.2 NP_001122.2 3 163
NM_001205220.1 NP_001192149.1 3 174
NM_170609.1 NP_733758.1 3 163
GenBank mRNA AB451334.1 - 2 84
AB451479.1 - 2 84
AK292497.1 - 3 144
BC131707.1 - 3 134
D38451.1 - 4 163
GQ472203.1 - 3 157
S80310.1 - 4 163
X95237.1 - 3 157
X95238.1 - 3 157

View Full Product Details