Gene Transcripts

Gene Symbol
BMX
Entrez Gene ID
660
Gene Name
BMX non-receptor tyrosine kinase
Gene Aliases
ETK, PSCTK2, PSCTK3
Chromosome Location
Chr. X: 15500777 - 15556529 on Build GRCh38
UniGene ID
Hs.495731
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320866.1 NP_001307795.1 17 1870
NM_001721.6 NP_001712.1 17 1811
NM_203281.2 NP_975010.1 17 1873
XM_017029752.1 XP_016885241.1 16 1626
GenBank mRNA AF045459.1 - 17 1884
AK291824.1 - 17 1843
AK313568.1 - 17 1806
BC016652.2 - 17 1828 3919253
U08341.1 - 2 95
X83107.1 - 17 1733

View Full Product Details