Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127384.2 | NP_001120856.1 | 2 | 233 | |
NM_001291133.1 | NP_001278062.1 | 3 | 310 | ||
NM_013266.3 | NP_037398.2 | 2 | 280 | ||
XM_017016151.1 | XP_016871640.1 | 2 | 206 | ||
XM_017016152.1 | XP_016871641.1 | 3 | 383 | ||
XM_017016153.1 | XP_016871642.1 | 3 | 283 | ||
XM_017016158.1 | XP_016871647.1 | 3 | 381 | ||
GenBank mRNA | AF091606.1 | - | 2 | 258 | |
AK289756.1 | - | 2 | 169 | ||
AK308726.1 | - | 2 | 147 | ||
AK313882.1 | - | 2 | 283 | ||
AY198183.1 | - | 3 | 249 | ||
BC022004.2 | - | 3 | 305 | ||
BC030646.2 | - | 3 | 305 | ||
BC065819.1 | - | 2 | 176 | 6189390 | |
BP362015.1 | - | 2 | 206 |