Gene Transcripts

Gene Symbol
AKAP8L
Entrez Gene ID
26993
Gene Name
A-kinase anchoring protein 8 like
Gene Aliases
HA95, HAP95, NAKAP, NAKAP95
Chromosome Location
Chr. 19: 15380048 - 15419121 on Build GRCh38
UniGene ID
Hs.399800
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291478.1 NP_001278407.1 9, 10 1180
NM_014371.3 NP_055186.3 9, 10 1363
XM_005259854.3 XP_005259911.2 10, 11 1168
XM_011527916.2 XP_011526218.1 9, 10 1270
GenBank mRNA AB025905.1 - 9, 10 1210
AB451326.1 - 8, 9 1165
AB451469.1 - 8, 9 1165
AF199414.1 - 8, 9 1165
AJ243467.1 - 9, 10 1211
AK225046.1 - 9, 10 1230
AK225291.1 - 9, 10 1230
AK295956.1 - 9, 10 1085
BC000713.2 - 9, 10 1215 3504388

View Full Product Details