Gene Transcripts

Gene Symbol
SCRN1
Entrez Gene ID
9805
Gene Name
secernin 1
Gene Aliases
SES1
Chromosome Location
Chr. 7: 29920103 - 29990289 on Build GRCh38
UniGene ID
Hs.520740
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145513.1 NP_001138985.1 8 1408
NM_001145514.1 NP_001138986.1 8 1340
NM_001145515.1 NP_001138987.1 7 1240
NM_014766.4 NP_055581.3 8 1400
GenBank mRNA AB071705.1 - 8 1290
AK289514.1 - 7 1435
AK294891.1 - 5 770
AK295172.1 - 8 1340
AK295713.1 - 7 1143
AK295800.1 - 5 759
BC040492.1 - 8 1352 5314711
D83777.2 - 8 1345

View Full Product Details