Gene Transcripts
- Gene Symbol
- VPS13C
- Entrez Gene ID
- 54832
- Gene Name
- vacuolar protein sorting 13 homolog C
- Gene Aliases
- PARK23
- Chromosome Location
- Chr. 15: 61852389 - 62060465 on Build GRCh38
- UniGene ID
- Hs.511668
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001018088.2 |
NP_001018098.1
|
10, 11 |
843 |
|
NM_017684.4 |
NP_060154.3
|
8, 9 |
714 |
|
NM_018080.3 |
NP_060550.2
|
8, 9 |
714 |
|
NM_020821.2 |
NP_065872.1
|
10, 11 |
843 |
|
XM_011521713.2 |
XP_011520015.1
|
10, 11 |
804 |
|
XM_011521714.2 |
XP_011520016.1
|
10, 11 |
804 |
|
GenBank mRNA |
AB037842.1 |
- |
10, 11 |
826 |
|
AJ608770.1 |
- |
8, 9 |
697 |
|
AJ608771.1 |
- |
10, 11 |
826 |
|
AJ626860.1 |
- |
8, 9 |
697 |
|
AJ626861.1 |
- |
10, 11 |
826 |
|
AK025364.1 |
- |
5, 6 |
365 |
|
AK307518.1 |
- |
8, 9 |
639 |
|
View Full Product Details