Gene Transcripts
- Gene Symbol
- FAM60A
- Entrez Gene ID
- 58516
- Gene Name
- family with sequence similarity 60 member A
- Gene Aliases
- C12orf14, L4, TERA
- Chromosome Location
- Chr. 12: 31280584 - 31326225 on Build GRCh38
- UniGene ID
- Hs.505154
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135811.1 |
NP_001129283.1
|
3, 4 |
453 |
|
NM_001135812.1 |
NP_001129284.1
|
2, 3 |
349 |
|
NM_021238.2 |
NP_067061.1
|
3, 4 |
449 |
|
XM_011520803.2 |
XP_011519105.1
|
2, 3 |
451 |
|
XM_017019758.1 |
XP_016875247.1
|
3, 4 |
517 |
|
XM_017019759.1 |
XP_016875248.1
|
4, 5 |
563 |
|
XM_017019760.1 |
XP_016875249.1
|
4, 5 |
568 |
|
XM_017019761.1 |
XP_016875250.1
|
3, 4 |
498 |
|
XM_017019762.1 |
XP_016875251.1
|
3, 4 |
415 |
|
GenBank mRNA |
AF087885.1 |
- |
3, 4 |
402 |
|
AF212220.1 |
- |
2, 3 |
308 |
|
AK026932.1 |
- |
2, 3 |
260 |
|
AK294457.1 |
- |
2, 3 |
362 |
|
AL157432.1 |
- |
3, 4 |
401 |
|
AY598323.1 |
- |
1, 2 |
127 |
|
BC000024.2 |
- |
3, 4 |
449 |
3504430 |
BC071966.1 |
- |
2, 3 |
229 |
5924274 |
BX648630.1 |
- |
1, 2 |
444 |
|
DA741767.1 |
- |
2, 3 |
496 |
|
View Full Product Details