Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289119.1 | NP_001276048.1 | 2 | 624 | |
NM_001289120.1 | NP_001276049.1 | 2 | 647 | ||
NM_001289121.1 | NP_001276050.1 | 2 | 765 | ||
NM_032226.2 | NP_115602.2 | 2 | 620 | ||
XM_005251608.4 | XP_005251665.1 | 2 | 1529 | ||
XM_005251612.2 | XP_005251669.1 | 2 | 587 | ||
XM_011518050.1 | XP_011516352.1 | 2 | 615 | ||
XM_017015197.1 | XP_016870686.1 | 2 | 742 | ||
XM_017015198.1 | XP_016870687.1 | 2 | 1228 | ||
XM_017015199.1 | XP_016870688.1 | 2 | 589 | ||
XM_017015200.1 | XP_016870689.1 | 2 | 862 | ||
GenBank mRNA | AK026264.1 | - | 2 | 510 | |
AK074608.1 | - | 1 | 109 | ||
AK315128.1 | - | 2 | 637 | ||
BC005990.1 | - | 2 | 553 | ||
BC022434.1 | - | 3 | 713 | ||
BC036940.1 | - | 2 | 738 | 5399765 |