Gene Transcripts

Gene Symbol
RASGEF1B
Entrez Gene ID
153020
Gene Name
RasGEF domain family member 1B
Gene Aliases
GPIG4
Chromosome Location
Chr. 4: 81426393 - 81471928 on Build GRCh38
UniGene ID
Hs.591696
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300735.1 NP_001287664.1 2 317
NM_001300736.1 NP_001287665.1 2 317
NM_152545.2 NP_689758.1 2 317
XM_017007812.1 XP_016863301.1 1 158
XM_017007813.1 XP_016863302.1 2 405
GenBank mRNA AK056257.1 - 2 296
AK291459.1 - 2 265
AK303625.1 - 2 293
BC036784.1 - 2 279
BC121003.1 - 2 232 40116919
BC121004.1 - 2 232 40116920
BX648337.1 - 2 230
DR002387.1 - 2 323

View Full Product Details