Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004459.6 | NP_004450.3 | 4 | 1761 | |
NM_182641.3 | NP_872579.2 | 4 | 1761 | ||
XM_005257150.3 | XP_005257207.1 | 4 | 1908 | ||
XM_005257151.3 | XP_005257208.1 | 4 | 1908 | ||
XM_005257152.2 | XP_005257209.1 | 4 | 1908 | ||
XM_005257153.3 | XP_005257210.1 | 4 | 1908 | ||
XM_005257154.3 | XP_005257211.1 | 4 | 1908 | ||
XM_005257155.3 | XP_005257212.1 | 4 | 1908 | ||
XM_005257156.3 | XP_005257213.1 | 4 | 1908 | ||
XM_005257157.3 | XP_005257214.1 | 4 | 1908 | ||
XM_005257158.3 | XP_005257215.1 | 4 | 1908 | ||
XM_005257159.2 | XP_005257216.1 | 4 | 1908 | ||
XM_005257160.2 | XP_005257217.1 | 4 | 1908 | ||
XM_005257161.3 | XP_005257218.1 | 4 | 1908 | ||
XM_011524520.2 | XP_011522822.1 | 4 | 1908 | ||
XM_011524521.2 | XP_011522823.1 | 4 | 1908 | ||
XM_011524522.2 | XP_011522824.1 | 4 | 1908 | ||
XM_011524523.2 | XP_011522825.1 | 4 | 1908 | ||
XM_011524525.2 | XP_011522827.1 | 4 | 1908 | ||
XM_011524526.2 | XP_011522828.1 | 4 | 1908 | ||
XM_017024353.1 | XP_016879842.1 | 4 | 1908 | ||
GenBank mRNA | AB032251.1 | - | 4 | 1754 | |
AK293224.1 | - | 4 | 1335 | ||
AK297281.1 | - | 5 | 1428 | ||
AY282495.1 | - | 4 | 1283 | ||
U05237.1 | - | 4 | 1356 |