Gene Transcripts

Gene Symbol
PIGH
Entrez Gene ID
5283
Gene Name
phosphatidylinositol glycan anchor biosynthesis class H
Gene Aliases
GPI-H
Chromosome Location
Chr. 14: 67589306 - 67600302 on Build GRCh38
UniGene ID
Hs.553497
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004569.3 NP_004560.1 2 366
XM_011536838.2 XP_011535140.1 2 364
XM_017021371.1 XP_016876860.1 2 368
XM_017021372.1 XP_016876861.1 2 359
GenBank mRNA AK293584.1 - 2 352
AK314108.1 - 2 333
BC004100.2 - 2 317 3936489
BC071849.1 - 2 314 5935599
BP244435.1 - 2 366
BT006804.1 - 2 269
L19783.1 - 2 329

View Full Product Details