Gene Transcripts

Gene Symbol
FBXL17
Entrez Gene ID
64839
Gene Name
F-box and leucine rich repeat protein 17
Gene Aliases
FBXO13, Fbl17, Fbx13
Chromosome Location
Chr. 5: 107859033 - 108382098 on Build GRCh38
UniGene ID
Hs.657225
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163315.2 NP_001156787.2 9 3205
GenBank mRNA AK126722.1 - 10 2393
AK172822.1 - 1 489

View Full Product Details