Gene Transcripts

Gene Symbol
PTCD2
Entrez Gene ID
79810
Gene Name
pentatricopeptide repeat domain 2
Gene Aliases
Chromosome Location
Chr. 5: 72320367 - 72368395 on Build GRCh38
UniGene ID
Hs.126906
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284403.1 NP_001271332.1 7 936
NM_001284404.1 NP_001271333.1 8 1066
NM_001284405.1 NP_001271334.1 9 1184
NM_024754.4 NP_079030.3 10 1263
XM_005248601.2 XP_005248658.1 9 1143
XM_005248603.2 XP_005248660.1 9 1119
XM_017009866.1 XP_016865355.1 9 1124
XM_017009867.1 XP_016865356.1 8 1036
XM_017009868.1 XP_016865357.1 8 1005
GenBank mRNA AK022660.1 - 8 1025
AK056761.1 - 10 1927
AK298750.1 - 7 927
AK303639.1 - 8 1035
BC018720.1 - 10 1239 4552964

View Full Product Details