Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032859.2 | NP_116248.2 | 2 | 551 | |
XM_011521128.2 | XP_011519430.1 | 2 | 509 | ||
GenBank mRNA | AK124864.1 | - | 1 | 953 | |
BC022566.1 | - | 2 | 467 | 4811611 | |
BQ424339.1 | - | 551 | |||
BX647428.1 | - | 1 | 131 |