Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145279.4 | NP_660322.2 | 4 | 1182 | |
NM_201403.2 | NP_958805.1 | 4 | 1200 | ||
XM_017000407.1 | XP_016855896.1 | 3 | 4131 | ||
GenBank mRNA | AK090453.1 | - | 3 | 4113 | |
AK091808.1 | - | 3 | 926 | ||
BC026078.2 | - | 4 | 1182 | ||
BC121169.2 | - | 4 | 1334 | 40122151 |