Gene Transcripts

Gene Symbol
UHRF2
Entrez Gene ID
115426
Gene Name
ubiquitin like with PHD and ring finger domains 2
Gene Aliases
NIRF, RNF107, TDRD23, URF2
Chromosome Location
Chr. 9: 6413148 - 6508108 on Build GRCh38
UniGene ID
Hs.493401
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152896.2 NP_690856.1 4 1029
NR_046386.1 4 1029
XM_011517704.2 XP_011516006.1 4 658
XM_011517705.2 XP_011516007.1 2 157
XM_017014254.1 XP_016869743.1 4 658
GenBank mRNA AB071698.1 - 4 727
AF274049.1 - 4 857
AK122730.1 - 2 131
AK308433.1 - 4 981
BC028397.2 - 4 1029 4797830

View Full Product Details