Gene Transcripts

Gene Symbol
ZC3H6
Entrez Gene ID
376940
Gene Name
zinc finger CCCH-type containing 6
Gene Aliases
ZC3HDC6
Chromosome Location
Chr. 2: 112275594 - 112340063 on Build GRCh38
UniGene ID
Hs.190477
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198581.2 NP_940983.2 9 1539
XM_006712519.2 XP_006712582.1 9 1546
GenBank mRNA AB111887.1 - 8 985
AK127776.1 - 6 608
AK131416.1 - 9 1525
BC155540.1 - 8 985
BC167154.1 - 8 985

View Full Product Details