Gene Transcripts
- Gene Symbol
- FBXO38
- Entrez Gene ID
- 81545
- Gene Name
- F-box protein 38
- Gene Aliases
- Fbx38, HMN2D, MOKA, SP329
- Chromosome Location
- Chr. 5: 148383935 - 148442836 on Build GRCh38
- UniGene ID
- Hs.483772
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271723.1 |
NP_001258652.1
|
11, 12 |
1583 |
|
NM_030793.4 |
NP_110420.3
|
11, 12 |
1583 |
|
XM_005268513.1 |
XP_005268570.1
|
11, 12 |
1572 |
|
XM_006714797.1 |
XP_006714860.1
|
11, 12 |
1572 |
|
XM_011537684.2 |
XP_011535986.1
|
11, 12 |
1625 |
|
XM_017009899.1 |
XP_016865388.1
|
12, 13 |
1720 |
|
XM_017009900.1 |
XP_016865389.1
|
12, 13 |
1784 |
|
XM_017009901.1 |
XP_016865390.1
|
11, 12 |
1637 |
|
XM_017009902.1 |
XP_016865391.1
|
11, 12 |
1625 |
|
GenBank mRNA |
AF251055.1 |
- |
4, 5 |
521 |
|
AK024024.1 |
- |
11, 12 |
1504 |
|
AK308414.1 |
- |
11, 12 |
1532 |
|
AL832841.1 |
- |
1, 2 |
50 |
|
BC005849.2 |
- |
1, 2 |
157 |
|
BC005873.2 |
- |
1, 2 |
157 |
|
BC033454.1 |
- |
11, 12 |
1516 |
4510274 |
BC050424.1 |
- |
10, 11 |
1488 |
6472693 |
View Full Product Details