Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004557.3 | NP_004548.3 | 5 | 969 | |
NR_134949.1 | 5 | 969 | |||
NR_134950.1 | 5 | 969 | |||
GenBank mRNA | AK290785.1 | - | 5 | 966 | |
BC063815.1 | - | 5 | 959 | ||
BC140782.1 | - | 5 | 924 | 9021650 | |
BC144540.1 | - | 5 | 930 | ||
BC144541.1 | - | 5 | 924 | ||
U95299.1 | - | 5 | 920 |