Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009013.3 | NP_033039.2 | 9 | 949 | |
GenBank mRNA | AK010920.1 | - | 7 | 948 | |
AK011379.1 | - | 9 | 950 | ||
AK077829.1 | - | 9 | 979 | ||
AK079525.1 | - | 9 | 971 | ||
BC003738.1 | - | 9 | 963 | 3495632 | |
BC022586.1 | - | 2 | 948 | ||
BC040809.1 | - | 2 | 948 | ||
U93583.1 | - | 9 | 947 |