Gene Transcripts
- Gene Symbol
- Rrh
- Entrez Gene ID
- 20132
- Gene Name
- retinal pigment epithelium derived rhodopsin homolog
- Gene Aliases
- Chromosome Location
- Chr. 3: 129808575 - 129822505 on Build GRCm38
- UniGene ID
- Mm.278758
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK044717.1 |
- |
|
1437 |
|
View Full Product Details