Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009270.3 | NP_033296.1 | 11 | 2385 | |
GenBank mRNA | AK138457.1 | - | 11 | 2398 | |
AK161837.1 | - | 10 | 2275 | ||
AK163502.1 | - | 11 | 2365 | ||
BC042781.1 | - | 11 | 1642 | 4915911 | |
BC056361.1 | - | 11 | 2385 | 6816800 | |
D42048.1 | - | 11 | 2251 |