Gene Transcripts

Gene Symbol
Ros1
Entrez Gene ID
19886
Gene Name
Ros1 proto-oncogene
Gene Aliases
Ros-1, c-ros
Chromosome Location
Chr. 10: 52044865 - 52195397 on Build GRCm38
UniGene ID
Mm.236163
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011282.2 NP_035412.2 5 590
GenBank mRNA U15443.1 - 5 589
X81650.1 - 5 413

View Full Product Details