Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253904.1 | NP_001240833.1 | 10 | 1109 | |
NM_001253905.1 | NP_001240834.1 | 6 | 663 | ||
NM_011424.3 | NP_035554.3 | 10 | 1274 | ||
GenBank mRNA | AF113002.1 | - | 6 | 655 | |
AF125671.1 | - | 10 | 1112 | ||
AK147394.1 | - | 10 | 1075 |