Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001113351.1 | NP_001106822.1 | 6 | 969 | |
NM_001113352.2 | NP_001106823.1 | 7 | 1058 | ||
NM_001113353.2 | NP_001106824.1 | 7 | 1058 | ||
NM_011523.2 | NP_035653.2 | 6 | 969 | ||
GenBank mRNA | AF041857.1 | - | 7 | 1059 | |
AF041862.1 | - | 6 | 914 | ||
AK014584.1 | - | 3 | 213 | ||
AK029755.1 | - | 4 | 476 | ||
AK038038.1 | - | 4 | 382 | ||
AK129122.1 | - | 5 | 486 | ||
AK142919.1 | - | 6 | 690 | ||
AK148053.1 | - | 7 | 797 | ||
AK163512.1 | - | 5 | 699 | ||
BC058749.1 | - | 7 | 935 | 6390688 | |
BC060214.1 | - | 6 | 815 | 6838657 |