Gene Transcripts

Gene Symbol
Slc18a3
Entrez Gene ID
20508
Gene Name
solute carrier family 18 (vesicular monoamine), member 3
Gene Aliases
VAChT, VAT
Chromosome Location
Chr. 14: 32462437 - 32464850 on Build GRCm38
UniGene ID
Mm.190503
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021712.2 NP_068358.2 1 859
GenBank mRNA AK158592.1 - 1 861
BC120498.1 - 1 476 40129421
BC137705.1 - 1 475 8860724

View Full Product Details