Gene Transcripts

Gene Symbol
Scx
Entrez Gene ID
20289
Gene Name
scleraxis
Gene Aliases
BB114693, Bhlha41, Scl
Chromosome Location
Chr. 15: 76456033 - 76459468 on Build GRCm38
UniGene ID
Mm.322821
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198885.3 NP_942588.1 2 824
GenBank mRNA AK013456.1 - 1 1700
BC062161.1 - 2 742 30138970
S78079.1 - 2 824

View Full Product Details