Gene Transcripts

Gene Symbol
Atxn7
Entrez Gene ID
246103
Gene Name
ataxin 7
Gene Aliases
A430107N12Rik, AI627028, Sca7, ataxin-7
Chromosome Location
Chr. 14: 13961439 - 14107302 on Build GRCm38
UniGene ID
Mm.133625
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139227.4 NP_631973.3 12 3548
GenBank mRNA AK046679.1 - 12 3549
BC035459.1 - 11 3201

View Full Product Details