Gene Transcripts

Gene Symbol
Dusp7
Entrez Gene ID
235584
Gene Name
dual specificity phosphatase 7
Gene Aliases
AU015694, Mkpx, Pyst2
Chromosome Location
Chr. 9: 106368610 - 106375724 on Build GRCm38
UniGene ID
Mm.275584
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153459.4 NP_703189.3 2 735
GenBank mRNA AK140372.1 - 2 736
BC010207.1 - 2 378 3964527

View Full Product Details