Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172300.3 | NP_758504.3 | 7 | 1465 | |
GenBank mRNA | AK005524.1 | - | 5 | 1016 | |
AK077793.1 | - | 7 | 1464 | ||
AK149770.1 | - | 7 | 1465 | ||
AK152529.1 | - | 7 | 1179 | ||
AK153328.1 | - | 7 | 1196 | ||
AK168942.1 | - | 7 | 1158 | ||
BC027266.1 | - | 7 | 1090 | ||
BC054412.1 | - | 7 | 1168 | 2646768 |