Gene Transcripts

Gene Symbol
Svs1
Entrez Gene ID
243377
Gene Name
seminal vesicle secretory protein 1
Gene Aliases
9530003A11
Chromosome Location
Chr. 6: 48986861 - 48991725 on Build GRCm38
UniGene ID
Mm.446302
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172888.3 NP_766476.2 1 770
GenBank mRNA AK035222.1 - 1 771
AY115568.1 - 1 727
AY283179.1 - 1 727
BC125635.1 - 1 641 40130150

View Full Product Details