Gene Transcripts

Gene Symbol
Lancl3
Entrez Gene ID
236285
Gene Name
LanC lantibiotic synthetase component C-like 3 (bacterial)
Gene Aliases
6030463G20Rik
Chromosome Location
Chr. X: 9199973 - 9268085 on Build GRCm38
UniGene ID
Mm.245119
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173414.3 NP_775590.2 4 1277
GenBank mRNA AK031623.1 - 4 1278
AK043841.1 - 4 478
AK045254.1 - 4 865
BC115944.1 - 3 400

View Full Product Details