Gene Transcripts

Gene Symbol
Fkrp
Entrez Gene ID
243853
Gene Name
fukutin related protein
Gene Aliases
A830029B19Rik, AI842067, AI847300, LGMD1I, MDC1C
Chromosome Location
Chr. 7: 16809267 - 16820668 on Build GRCm38
UniGene ID
Mm.39703
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173430.2 NP_775606.1 3 2472
GenBank mRNA AJ511806.1 - 3 2455
AK080624.1 - 1 2282
AK135089.1 - 3 2472
AK156156.1 - 3 2471
AK168308.1 - 3 2470
AK209876.1 - 1 320
BC053072.1 - 3 2455 5686286

View Full Product Details