Gene Transcripts

Gene Symbol
Aard
Entrez Gene ID
239435
Gene Name
alanine and arginine rich domain containing protein
Gene Aliases
A5D3, AV328152
Chromosome Location
Chr. 15: 52040107 - 52045722 on Build GRCm38
UniGene ID
Mm.24204
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175503.3 NP_780712.2 2 1090
GenBank mRNA AK046414.1 - 2 1091
AY134665.1 - 2 1029
BC089505.1 - 2 1044 6772439

View Full Product Details