Gene Transcripts

Gene Symbol
RAB33A
Entrez Gene ID
9363
Gene Name
RAB33A, member RAS oncogene family
Gene Aliases
RabS10
Chromosome Location
Chr. X: 130110633 - 130184872 on Build GRCh38
UniGene ID
Hs.654356
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004794.2 NP_004785.1 2 953
XM_017029963.1 XP_016885452.1 3 642
GenBank mRNA AF498959.1 - 2 689
AK094927.1 - 3 1631
BC001157.1 - 2 953 3510745
BC009996.2 - 2 953 4129324
BT007038.1 - 2 689
D14889.1 - 2 1126

View Full Product Details