Gene Transcripts

Gene Symbol
Prr11
Entrez Gene ID
270906
Gene Name
proline rich 11
Gene Aliases
B930067F20Rik
Chromosome Location
Chr. 11: 87089156 - 87108714 on Build GRCm38
UniGene ID
Mm.132381
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175563.5 NP_780772.1 3 276
GenBank mRNA AK040348.1 - 3 254
AK047463.1 - 3 254
AK083651.1 - 3 271
BC023918.1 - 3 286 5323297

View Full Product Details