Gene Transcripts

Gene Symbol
Kctd12
Entrez Gene ID
239217
Gene Name
potassium channel tetramerisation domain containing 12
Gene Aliases
AU046135, AW538430, Pfet1, Pfetin
Chromosome Location
Chr. 14: 102976581 - 102982637 on Build GRCm38
UniGene ID
Mm.246466
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177715.4 NP_808383.3 1 1158
GenBank mRNA AK035535.1 - 1 1160
AK087895.1 - 1 1166
AK172581.1 - 1 1159
AK173260.1 - 1 663
BC116647.1 - 2 545
BC116648.1 - 2 545

View Full Product Details