Gene Transcripts

Gene Symbol
Pcid2
Entrez Gene ID
234069
Gene Name
PCI domain containing 2
Gene Aliases
A730042J05Rik
Chromosome Location
Chr. 8: 13077183 - 13105343 on Build GRCm38
UniGene ID
Mm.103427
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK082891.1 - 14 2495
BC072621.1 - 11 2358
BC118533.1 - 14 2424 40095328

View Full Product Details